Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human STAT5a/b Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC41901N100UG
This item is not returnable.
View return policy
Description
STAT5a/b Monoclonal specifically detects STAT5a/b in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
STAT5a/b | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
MGF, STAT5, STAT5A signal transducer and activator of transcription 5A | |
Phosphopeptide containing human STAT5b Y699 site (amino acid sequence of this peptide is identical to a corresponding region of human STAT5a containing Y694), Accession # P51692 | |
100 μg | |
Primary | |
Detects human STAT5a/b when phosphorylated at Y964/Y699 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Flow Cytometry | |
1247C | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P51692 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
6776 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction