Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human STAT5b Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC1584N100UG
This item is not returnable.
View return policy
Description
STAT5b Monoclonal specifically detects STAT5b in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
STAT5b | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_036580 | |
STAT5B | |
Human STAT5b synthetic peptide MDSQWIPHAQS Accession # NP_036580 | |
RUO | |
6777 | |
Human | |
IgG2b |
Flow Cytometry | |
389215 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
Mouse | |
100 μg | |
Primary | |
Detects human STAT5b in Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction