Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human STIM1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB69761
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB69761 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB69761 Supplier R&D Systems Supplier No. MAB69761
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

STIM1 Monoclonal specifically detects STIM1 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen STIM1
Applications Immunohistochemistry
Classification Monoclonal
Clone 705129
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q13586
Gene Alias GOK, GOKD11S4896E, stromal interaction molecule 1
Gene Symbols STIM1
Host Species Mouse
Immunogen E. coli-derived recombinant human STIM1 Leu23-Thr182 Accession # Q13586
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6786
Test Specificity Detects human STIM1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human STIM2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.