Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human STING/TMEM173 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC7169U100UG
This item is not returnable.
View return policy
Description
STING/TMEM173 Monoclonal specifically detects STING/TMEM173 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
STING/TMEM173 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q86WV6 | |
TMEM173 | |
E. coli-derived recombinant human STING/TMEM173 Ala215-Ser379 Accession # Q86WV6 | |
RUO | |
340061 | |
Human | |
IgG2b |
Flow Cytometry | |
723505 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, Stimulator of interferon genes protein, sting, STINGhSTING, TMEM173, transmembrane protein 173 | |
Mouse | |
100 μg | |
Primary | |
Detects human STING/TMEM173 in direct ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction