Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human TNF RII/TNFRSF1B Quantikine ELISA Kit

Catalog No. p-4753700
Click to view available options
Quantity:
1 Plate
6 Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Human sTNF RII/TNFRSF1B Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human sTNF RII/TNFRSF1B Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human TNF RII/TNFRSF1B in cell culture supernates (200 ul), serum (20 ul), edta plasma (20 ul), heparin plasma (20 ul), citrate plasma (20 ul), urine (20 ul).

Specifications

Assay Range 7.80 - 500pg/mL
Assay Sensitivity 2.3pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine
Specificity Natural and Recombinant Human sTNF RII
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 7133
Quantity 1 Plate
Sample Volume Cell Culture Supernates (200uL),Serum (20uL),EDTA Plasma (20uL),Heparin Plasma (20uL),Citrate Plasma (20uL),Urine (20uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNFRII, TNF-RII, TNF-R-II, TNFR-II, TNFRSF1B, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1 B, tumor necrosis factor receptor superfamily, member 1 B, Tumor necrosis factor receptor type II
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.