Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human STRA6 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC5904G100UG
This item is not returnable.
View return policy
Description
STRA6 Monoclonal specifically detects STRA6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
STRA6 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
AAH25256 | |
STRA6 | |
NS0 mouse myeloma cell line transfected with human STRA6 Met1-Pro667 Accession # AAH25256 | |
RUO | |
64220 | |
Human | |
IgG1 |
Flow Cytometry | |
496613 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
FLJ12541, MCOPS9, stimulated by retinoic acid gene 6 homolog (mouse), stimulated by retinoic acid gene 6 protein homolog | |
Mouse | |
100 μg | |
Primary | |
Detects human STRA6 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction