Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human STRADB Partial ORF (AAH08302, 319 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7101044
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein that belongs to the serine/threonine protein kinase STE20 subfamily. One of the active site residues in the protein kinase domain of this protein is altered, and it is thus a pseudokinase. This protein is a component of a complex involved in the activation of serine/threonine kinase 11, a master kinase that regulates cell polarity and energy-generating metabolism. This complex regulates the relocation of this kinase from the nucleus to the cytoplasm, and it is essential for G1 cell cycle arrest mediated by this kinase. The protein encoded by this gene can also interact with the X chromosome-linked inhibitor of apoptosis protein, and this interaction enhances the anti-apoptotic activity of this protein via the JNK1 signal transduction pathway. Two pseudogenes, located on chromosomes 1 and 7, have been found for this gene. [provided by RefSeq]

Sequence: SSGTHTVNSDRLHTPSSKTFSPAFFSLVQLCLQQDPEKRPSASSLLSHVFFKQMKEESQDSILSLLPPAYNKPSISLPPVLPWTEPECDFPDEKDSYWEF

Specifications

Accession Number AAH08302
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55437
Molecular Weight (g/mol) 36.63kDa
Name STRADB (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen SSGTHTVNSDRLHTPSSKTFSPAFFSLVQLCLQQDPEKRPSASSLLSHVFFKQMKEESQDSILSLLPPAYNKPSISLPPVLPWTEPECDFPDEKDSYWEF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ALS2CR2/CALS-21/ILPIP/ILPIPA/MGC102916/PAPK/PRO1038
Common Name STRADB
Gene Symbol STRADB
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.