Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Sulfatase Modifying Factor 1/SUMF1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3680
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3680 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3680 Supplier R&D Systems Supplier No. AF3680
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Sulfatase Modifying Factor 1/SUMF1 Polyclonal specifically detects Sulfatase Modifying Factor 1/SUMF1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Sulfatase Modifying Factor 1/SUMF1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q8NBK3
Gene Alias AAPA3037, EC 1.8.99.-, FGE1, FGEC-alpha-formylglycine-generating enzyme 1, FGly-generating enzyme, MGC131853, MGC150436, sulfatase modifying factor 1, sulfatase-modifying factor 1, UNQ3037
Gene Symbols SUMF1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human SUMF1 isoform 1 Ser34-Asp374 with a Ser63Asn substitution Accession # Q8NBK3
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285362
Test Specificity Detects human SUMF1 in direct ELISAs and Western blots. In these formats, approximately 45% cross-reactivity with recombinant mouse SUMF1 is observed and less than 1% cross-reactivity with recombinant human SUMF2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.