Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Sulfatase Modifying Factor 2/SUMF2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3594
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3594 100 μg
MAB3594SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3594 Supplier R&D Systems Supplier No. MAB3594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sulfatase Modifying Factor 2/SUMF2 Monoclonal specifically detects Sulfatase Modifying Factor 2/SUMF2 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Sulfatase Modifying Factor 2/SUMF2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 330312
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8NBJ7
Gene Alias C-alpha-formyglycine-generating enzyme 2, C-alpha-formylglycine-generating enzyme 2, DKFZp566I1024, DKFZp686I1024, DKFZp686L17160, DKFZp781L1035, FGE2, MGC99485, paralog of the formylglycine-generating enzyme, pFGE, sulfatase modifying factor 2, sulfatase-modifying factor 2
Gene Symbols SUMF2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Sulfatase Modifying Factor 2/SUMF2 Gln26-Leu301 Accession # Q8NBJ7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25870
Test Specificity Detects human Sulfatase Modifying Factor 2/SUMF2 in direct ELISAs and Western blots. No cross-reactivity with recombinant human SUMF1 or recombinant mouse SUMF2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.