Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SUMO3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2959R100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2959R100 100 μg
MAB2959RSP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2959R100 Supplier R&D Systems Supplier No. MAB2959R100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

SUMO3 Monoclonal specifically detects SUMO3 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen SUMO3
Applications Immunocytochemistry
Classification Monoclonal
Clone 401513R
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P55854
Gene Alias SMT3 (suppressor of mif two 3, yeast) homolog 1, SMT3 homolog 1, SMT3 suppressor of mif two 3 homolog 1, SMT3 suppressor of mif two 3 homolog 3 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 3 (yeast), SMT3A, SMT3B, SMT3H1, SMT3H1small ubiquitin-related modifier 3, SUMO-2, SUMO-3, ubiquitin-like protein SMT3A, Ubiquitin-like protein SMT3B
Gene Symbols SUMO3
Host Species Rat
Immunogen E. coli-derived recombinant human SUMO3 Met1-Phe103 Accession # P55854
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6612
Test Specificity Detects human SUMO3 in direct ELISAs.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.