Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SUMO3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB2959
This item is not returnable.
View return policy
Description
SUMO3 Monoclonal specifically detects SUMO3 in Human samples. It is validated for Immunocytochemistry.Specifications
SUMO3 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
P55854 | |
SUMO3 | |
E. coli-derived recombinant human SUMO3 Met1-Phe103 Accession # P55854 | |
100 μg | |
Primary | |
Detects human SUMO3 in direct ELISAs. | |
Human | |
Purified |
Immunocytochemistry | |
401513 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
SMT3 (suppressor of mif two 3, yeast) homolog 1, SMT3 homolog 1, SMT3 suppressor of mif two 3 homolog 1, SMT3 suppressor of mif two 3 homolog 3 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 3 (yeast), SMT3A, SMT3B, SMT3H1, SMT3H1small ubiquitin-related modifier 3, SUMO-2, SUMO-3, ubiquitin-like protein SMT3A, Ubiquitin-like protein SMT3B | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6612 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction