Learn More
Human Survivin Affinity Purified Polyclonal Ab, R&D Systems™

Description
Specifications
Specifications
| Antigen | Survivin |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 1-15 ug/mL, CyTOF-ready |
| Gene Alias | API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha |
| Gene Symbols | BIRC5 |
| Host Species | Goat |
| Immunogen | E. coli-derived recombinant human Survivin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.