Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SUSD2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Catalog No. FB90561R025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB90561R025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB90561R025 Supplier R&D Systems Supplier No. FAB90561R025
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SUSD2 Monoclonal specifically detects SUSD2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SUSD2
Applications Flow Cytometry
Classification Monoclonal
Clone 944827
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 5 μL/106 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9UGT4
Gene Alias BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived human SUSD2, Met1-Ala785, Accession No. Q9UGT4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Primary or Secondary Primary
Gene ID (Entrez) 56241.0
Test Specificity Detects human SUSD2 in direct ELISAs. Stains human SUSD2 transfectants but not irrelevant transfectants in flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.