Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SUSD2 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB90561N100UG
This item is not returnable.
View return policy
Description
SUSD2 Monoclonal specifically detects SUSD2 in Human samples. It is validated for Flow Cytometry.Specifications
SUSD2 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2 | |
Human embryonic kidney cell line HEK293-derived human SUSD2 Met1-Ala785 Accession # Q9UGT4 | |
RUO | |
56241 | |
Human | |
IgG2a |
Flow Cytometry | |
944827 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9UGT4 | |
Mouse | |
100 μg | |
Primary | |
Detects human SUSD2 in direct ELISAs. Stains human SUSD2 transfectants but not irrelevant transfectants in flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction