Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Synaptophysin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5555SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5555SP 25 μg
MAB5555100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5555SP Supplier R&D Systems Supplier No. MAB5555SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Synaptophysin Monoclonal specifically detects Synaptophysin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen Synaptophysin
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 959904
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P08247
Gene Alias Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin, SYP
Host Species Mouse
Immunogen E.coli-derived recombinant human Synaptophysin Glu50-Phe106 Accession # P08247
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6855
Test Specificity Detects human Synaptophysin in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.