Learn More
Human Syndecan-1/CD138 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Syndecan-1/CD138 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 359103R |
| Conjugate | Alexa Fluor 350 |
| Dilution | Flow Cytometry 0.25-1 μg/106 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | NP_002988 |
| Gene Alias | CD138, CD138 antigen, SDC, SDC1, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1, Syndecan1 |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.