Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Syndecan-2/CD362 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2965P100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2965P100 100 Tests
FAB2965P025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB2965P100 Supplier R&D Systems Supplier No. FAB2965P100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Syndecan-2/CD362 Monoclonal specifically detects Syndecan-2/CD362 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Syndecan-2/CD362
Applications Flow Cytometry
Classification Monoclonal
Clone 305515
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. AAH49836.1
Gene Alias CD362, CD362 antigen, Fibroglycan, heparan sulfate proteoglycan 1, cell surface-associated, Heparan sulfate proteoglycan core protein, HSPG, HSPG1, HSPG1syndecan proteoglycan 2, SDC2, SYND2, SYND2cell surface-associated heparan sulfate proteoglycan 1, syndecan 2, syndecan-2, Syndecan2
Gene Symbols SDC2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Syndecan-2 Glu19-Gly144 Accession # AAH49836.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6383
Test Specificity Detects human Syndecan-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombiant mouse Syndecan-1, recombinant human (rh) Syndecan-3, or rhSyndecan-4 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.