Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Syndecan-3 Isoform 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3539
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3539 100 μg
AF3539SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3539 Supplier R&D Systems Supplier No. AF3539
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

SDC3 Polyclonal specifically detects SDC3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen SDC3
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
Gene Accession No. O75056
Gene Alias KIAA0468, N-syndecan, SDC3, SDCN, SYND3syndecan proteoglycan 3, syndecan 3, syndecan 3 (N-syndecan), syndecan neural type, syndecan-3, Syndecan3
Gene Symbols SDC3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Syndecan-3 Gln48-Lys383 Accession # O75056
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9672
Test Specificity Detects human Syndecan-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse Syndecan-3 is observed and less than 1% cross-reactivity with recombinant mouse Syndecan-4 and recombinant human Syndecan-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.