Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Syntaxin 6 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF5664
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF5664 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF5664 Supplier R&D Systems Supplier No. BAF5664
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Syntaxin 6 Polyclonal specifically detects Syntaxin 6 in Human samples. It is validated for Immunocytochemistry.

Order Info

The product is shipped at ambient temperature.

Specifications

Antigen Syntaxin 6
Applications Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Accession No. O43752
Gene Alias STX6, syntaxin 6, syntaxin-6
Gene Symbols STX6
Host Species Sheep
Immunogen E. coli-derived recombinant human Syntaxin 6 Ser69-Asn217 Accession # O43752
Purification Method Antigen Affinity-purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10228
Test Specificity Syntaxin 6 antibodies are ideal for immunocytochemistry colocalization studies in trans-Golgi and endosomal membranes. The unconjugated antibody detects human Syntaxin 6 in direct ELISAs and Western blots. In direct ELISAs, less than 3% cross-reactivity with recombinant human (rh) Syntaxin 8, rhSyntaxin 12, and rhSyntaxin 16 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.