Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Syntaxin-BP3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5659SP
This item is not returnable.
View return policy
Description
Syntaxin-BP3/Munc18c Polyclonal specifically detects Syntaxin-BP3/Munc18c in Human samples. It is validated for Western Blot.Specifications
Syntaxin-BP3/Munc18c | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MUNC18-3, Munc18c, Platelet Sec1 protein, Protein unc-18 homolog 3, Protein unc-18 homolog C, PSP, STXBP3, syntaxin 4 binding protein, syntaxin binding protein 3, syntaxin-binding protein 3, SyntaxinBP3, unc18-3, Unc-18C | |
Goat | |
Affinity Purified | |
RUO | |
6814 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
O00186 | |
STXBP3 | |
E. coli-derived recombinant human Syntaxin-BP3 Thr84-Thr231 Accession # O00186 | |
25 μg | |
Primary | |
Detects human Syntaxin-BP3 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) STXBP1 and rhSTXBP2 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction