Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Syntaxin-BP3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5659
This item is not returnable.
View return policy
Description
Syntaxin-BP3/Munc18c Polyclonal specifically detects Syntaxin-BP3/Munc18c in Human samples. It is validated for Western Blot.Specifications
Syntaxin-BP3/Munc18c | |
Polyclonal | |
Unconjugated | |
O00186 | |
STXBP3 | |
E. coli-derived recombinant human Syntaxin-BP3 Thr84-Thr231 Accession # O00186 | |
100 μg | |
Primary | |
Detects human Syntaxin-BP3 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) STXBP1 and rhSTXBP2 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
MUNC18-3, Munc18c, Platelet Sec1 protein, Protein unc-18 homolog 3, Protein unc-18 homolog C, PSP, STXBP3, syntaxin 4 binding protein, syntaxin binding protein 3, syntaxin-binding protein 3, SyntaxinBP3, unc18-3, Unc-18C | |
Goat | |
Affinity Purified | |
RUO | |
6814 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction