Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human T-bet/TBX21 Fluorescein-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems IC5385F
This item is not returnable.
View return policy
Description
T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
T-bet/TBX21 | |
Monoclonal | |
Fluorescein | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Tbet, T-bet, TBETT-PET, TBLYM, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, TBX21, T-PET, Transcription factor TBLYM | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
30009 | |
Human | |
Purified |
Flow Cytometry | |
525803 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
Q9UL17 | |
TBX21 | |
E. coli-derived recombinant human T-bet Glu326-Asn535 Accession # Q9UL17 | |
100 Tests | |
Primary | |
Detects human T-bet. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction