Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human T-bet/TBX21 PerCP-conjugated Antibody, R&D Systems™

Catalog No. IC5385C
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5385C 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC5385C Supplier R&D Systems Supplier No. IC5385C
This item is not returnable. View return policy

Mouse Monoclonal Antibody

T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen T-bet/TBX21
Applications Flow Cytometry
Classification Monoclonal
Clone 525803
Conjugate PerCP
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9UL17
Gene Alias Tbet, T-bet, TBETT-PET, TBLYM, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, TBX21, T-PET, Transcription factor TBLYM
Gene Symbols TBX21
Host Species Mouse
Immunogen E. coli-derived recombinant human T-bet Glu326-Asn535 Accession # Q9UL17
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30009
Test Specificity Detects human T-bet.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.