Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human t-Plasminogen Activator/tPA ELISA Kit (Colorimetric)
SDP

Catalog No. NBP191260 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP191260 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP191260 Supplier Novus Biologicals™ Supplier No. NBP191260

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human t-Plasminogen Activator/tPA ELISA Kit (Colorimetric) measures human t-Plasminogen Activator/tPA in cell culture supernatant, serum.

Specifications

Assay Range 15.6 - 1,000pg/mL
Assay Sensitivity 6pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 5327
Gene Symbol PLAT
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with polyclonal antibody to human t-PA, HRP-Conjugate anti-human t-PA polyclonal antibody, Human t-PA Standard lyophilized, 2ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Storage Requirements Store at 4°C. Do not freeze.
Synonym alteplase, DKFZp686I03148, EC 3.4.21, EC 3.4.21.68, plasminogen activator, tissue, plasminogen activator, tissue type, reteplase, tissue plasminogen activator (t-PA), tissue-type plasminogen activator, t-PA, TPA, t-plasminogen activator
Target t-Plasminogen Activator/tPA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.