Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TACE/ADAM17 Cytosolic Domain Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2129
This item is not returnable.
View return policy
Description
TACE/ADAM17 Polyclonal specifically detects TACE/ADAM17 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Knockout Validated.Specifications
TACE/ADAM17 | |
Polyclonal | |
Unconjugated | |
P78536 | |
ADAM17 | |
E. coli-derived recombinant human TACE Asp695-Cys824 Accession # P78536 | |
100 μg | |
Primary | |
Detects human TACE in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) BACE, rhADAM8, rhADAM9, rhADAM10, and rhADAM15 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, KnockDown | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Knockout Validated | |
ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM17, ADAM18, CD156b, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme | |
Goat | |
Affinity Purified | |
RUO | |
6868 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction