Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TACE/ADAM17 Ectodomain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9304
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9304 500 μg
MAB9304SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9304 Supplier R&D Systems Supplier No. MAB9304
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

TACE/ADAM17 Monoclonal specifically detects TACE/ADAM17 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen TACE/ADAM17
Applications ELISA
Classification Monoclonal
Clone 111608
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P78536
Gene Alias ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM17, ADAM18, CD156b, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme
Gene Symbols ADAM17
Host Species Mouse
Immunogen Insect ovarian cell line T. ni-derived recombinant human TACE/ADAM17 Pro18-Asn671 (predicted) Accession # P78536
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6868
Test Specificity Detects human TACE/ADAM17 Ectodomain in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant mouse (rm) TACE, recombinant human (rh) ADAM8, rhADAM9, or rmADAM10 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.