Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TAF5L Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6785
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6785 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6785 Supplier R&D Systems Supplier No. AF6785
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

TAF5L Polyclonal specifically detects TAF5L in Human samples. It is validated for Western Blot.

Specifications

Antigen TAF5L
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. O75529
Gene Alias PAF65B, PAF65BPAF65-beta, PCAF associated factor 65 beta, PCAF-associated factor 65 beta, TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDasubunit 5L, TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65 kD, TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65kDa
Gene Symbols TAF5L
Host Species Sheep
Immunogen E. coli-derived recombinant human TAF5L Met1-Tyr189 Accession # O75529
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27097
Test Specificity Detects human TAF5L in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human TAF1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.