Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TAFA2/FAM19A2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4179
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4179 100 μg
MAB4179SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4179 Supplier R&D Systems Supplier No. MAB4179
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TAFA2/FAM19A2 Monoclonal specifically detects TAFA2/FAM19A2 in Human samples. It is validated for Western Blot.

Specifications

Antigen TAFA2/FAM19A2
Applications Western Blot
Classification Monoclonal
Clone 447815
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH28403
Gene Alias Chemokine-like protein TAFA-2, DKFZp761E1217, DKFZp781P0552, FAM19A2, family with sequence similarity 19 (chemokine (C-C motif)-like), member A2, MGC42403, TAFA2, TAFA-2
Gene Symbols FAM19A2
Host Species Mouse
Immunogen E. coli-derived recombinant human TAFA2/FAM19A2 Ala31-His131 Accession # AAH28403
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338811
Test Specificity Detects human TAFA2/FAM19A2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA1, 3, 4, or 5 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.