Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TAFA2/FAM19A2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4179
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4179 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4179 Supplier R&D Systems Supplier No. AF4179
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

TAFA2/FAM19A2 Polyclonal specifically detects TAFA2/FAM19A2 in Human samples. It is validated for Immunohistochemistry, Neutralization.

Specifications

Antigen TAFA2/FAM19A2
Applications Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL, Neutralization 5 ug/mL
Gene Accession No. Q8N3H0
Gene Alias Chemokine-like protein TAFA-2, DKFZp761E1217, DKFZp781P0552, FAM19A2, family with sequence similarity 19 (chemokine (C-C motif)-like), member A2, MGC42403, TAFA2, TAFA-2
Gene Symbols FAM19A2
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human TAFA2/FAM19A2 Ala31-His131 Accession # Q8N3H0
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338811
Test Specificity Detects human TAFA2/FAM19A2 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human TAFA5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.