Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TAFA4/FAM19A4 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC5099V100UG
This item is not returnable.
View return policy
Description
TAFA4/FAM19A4 Monoclonal specifically detects TAFA4/FAM19A4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
TAFA4/FAM19A4 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q96LR4 | |
FAM19A4 | |
E. coli-derived recombinant human TAFA4 Ser35-Arg140 Accession # Q96LR4 | |
RUO | |
151647 | |
Human | |
IgG1 |
Flow Cytometry | |
480103 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Chemokine-like protein TAFA-4, FAM19A4, family with sequence similarity 19 (chemokine (C-C motif)-like), member A4, FLJ25161, TAFA4, TAFA-4 | |
Mouse | |
100 μg | |
Primary | |
Detects human TAFA4/FAM19A4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA1, 2, 3, or 5 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction