Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TAFA5/FAM19A5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5148SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5148SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5148SP Supplier R&D Systems Supplier No. MAB5148SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TAFA5/FAM19A5 Monoclonal specifically detects TAFA5/FAM19A5 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TAFA5/FAM19A5
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 463102
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_056196
Gene Alias Chemokine-like protein TAFA-5, FAM19A5, family with sequence similarity 19 (chemokine (C-C motif)-like), member A5, QLLK5208, TAFA protein 5, TAFA5, TAFA-5, UNQ5208
Gene Symbols FAM19A5
Host Species Rat
Immunogen E. coli-derived recombinant human TAFA5/FAM19A5 Gln26-Ser125 Accession # NP_056196
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25817
Test Specificity Detects TAFA5/FAM19A5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA2, 3, or 4 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.