Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Talin2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4738SP
This item is not returnable.
View return policy
Description
Talin2 Polyclonal specifically detects Talin2 in Human samples. It is validated for Western Blot.Specifications
Talin2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp451B1011, DKFZp686I0976, DKFZp686K0979, ILWEQ, KIAA0320, talin2, talin-2, TLN 2, TLN2 | |
Goat | |
Affinity Purified | |
RUO | |
83660 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9Y4G6 | |
TLN2 | |
E. coli-derived recombinant human Talin2 Ala977-Arg1183 Accession # Q9Y4G6 | |
25 μg | |
Primary | |
Detects endogenous human Talin2 in Western blots. In Western blots, no cross-reactivity to recombinant human Talin1 was detected. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction