Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TAOK3 (NM_016281.2, 1 a.a. - 898 a.a.) Full-length Recombinant Protein expressed in Sf9 cells

Catalog No. 89964050 Shop All Abnova Corporation Products
Click to view available options
Quantity:
100 μg

Used for Func, SDS-PAGE

Specifications

Accession Number NM_016281.2
Concentration 0.124 ug/uL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 51347
Molecular Weight (g/mol) 106.015kDa
Name TAOK3 (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Purification Method GST affinity chromatography
Quality Control Testing 2 ug/lane SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing
Regulatory Status RUO
Gene Alias DKFZp666H245/DPK/FLJ31808/JIK/MAP3K18
Common Name TAOK3
Gene Symbol TAOK3
Biological Activity 1pmol/μg x min
Species Baculovirus
Recombinant Recombinant
Protein Tag None
Expression System Insect cell (Sf9) expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.