Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TAP2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | TAP2 |
---|---|
Clone | 998524 |
Dilution | Western Blot 2 μg/mL, Immunohistochemistry 8-25 μg/mL, Immunocytochemistry 8-25 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2432SP
![]() |
R&D Systems
MAB2432SP |
25 μg |
Each for $200.50
|
|
|||||
MAB2432100
![]() |
R&D Systems
MAB2432100 |
100 μg |
Each for $467.50
|
|
|||||
Description
TAP2 Monoclonal specifically detects TAP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
TAP2 | |
Western Blot 2 μg/mL, Immunohistochemistry 8-25 μg/mL, Immunocytochemistry 8-25 μg/mL | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ABC18, ABCB3, ABCB3ABC18, antigen peptide transporter 2, APT2, ATP-binding cassette, sub-family B (MDR/TAP), member 3, D6S217E, MHC 2, Peptide transporter involved in antigen processing 2, Peptide transporter PSF2, Peptide transporter TAP2, PSF2, PSF2PSF-2, Really interesting new gene 11 protein, RING11, RING11ATP-binding cassette sub-family B member 3, transporter 2, ABC (ATP binding cassette), transporter 2, ATP-binding cassette, sub-family B (MDR/TAP), Y1 | |
Synthetic peptide containing human TAP2, Accession # Q03519 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TAP2 in direct ELISAs. |
998524 | |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Human | |
Q03519 | |
6891 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title