Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Tau MAb (Clone 376720), R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3494SP
This item is not returnable.
View return policy
Description
Tau Monoclonal specifically detects Tau in Multi-Species samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Tau | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
P10636 | |
MAPT | |
E. coli-derived recombinant human Tau Gln624-Gln756 Accession # P10636 | |
25 μg | |
Primary | |
Detects human Tau in direct ELISAs and Western blots. | |
Multi-species | |
Purified |
Western Blot, Immunohistochemistry | |
376720 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MAPT, MGC138549, microtubule-associated protein tau, MSTD, MSTDMAPTL, MTBT1, MTBT1Neurofibrillary tangle protein, MTBT2, Neurofibrillary tangle protein, PHF-tau, PPND, tau, TAUPaired helical filament-tau | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4137 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction