Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TBK1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF9934SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF9934SP 25 μg
AF9934100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF9934SP Supplier R&D Systems Supplier No. AF9934SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Antibody

TBK1 specifically detects TBK1 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen TBK1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Simple Western 20 ug/mL, Immunohistochemistry 3-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9UHD2
Gene Alias EC 2.7.11, EC 2.7.11.1, FLJ11330, FTDALS4, NAK, NAK serine/threonine-protein kinase TBK 1, NAKserine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, NFKB-Activating Kinase, T2K, TANK-binding kinase 1NF-kB-activating kinase, TBK 1
Host Species Sheep
Immunogen E. coli-derived recombinant human TBK-1, Met1-Gly121, Accession No. Q9UHD2
Purification Method Antigen Affinity-purified
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 29110
Test Specificity Detects human TBK-1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.