Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TBX18 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6337SP
This item is not returnable.
View return policy
Description
TBX18 Monoclonal specifically detects TBX18 in Human samples. It is validated for Western Blot.Specifications
TBX18 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL | |
O95935 | |
TBX18 | |
E. coli-derived recombinant human TBX18 Ala384-Ser504 Accession # O95935 | |
25 μg | |
Primary | |
Detects human TBX18 in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
635303 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
T-box 18, T-box protein 18, T-box transcription factor TBX18 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9096 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction