Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TBX2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB50401SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB50401SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB50401SP Supplier R&D Systems Supplier No. MAB50401SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TBX2 Monoclonal specifically detects TBX2 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen TBX2
Applications Immunocytochemistry
Classification Monoclonal
Clone 393720
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q13207
Gene Alias FLJ10169, T-box 2, T-box protein 2, T-box transcription factor TBX2
Gene Symbols TBX2
Host Species Rat
Immunogen E. coli-derived recombinant human TBX2 Thr593-Arg702 Accession # Q13207
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6909
Test Specificity Detects human TBX2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TBX3, rhTBX6, or rhT-bet is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.