Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TBX20 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8124
This item is not returnable.
View return policy
Description
TBX20 Monoclonal specifically detects TBX20 in Human samples. It is validated for Immunocytochemistry.Specifications
TBX20 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
Q9UMR3 | |
TBX20 | |
E. coli-derived recombinant human TBX20 Met1-Leu116 Accession # Q9UMR3 | |
100 μg | |
Primary | |
Detects human TBX20 in ELISA. | |
Human | |
Purified |
Immunocytochemistry | |
668710 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ASD4, T-box 20, T-box protein 20, T-box transcription factor TBX20 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
57057 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction