Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TBX3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4509
This item is not returnable.
View return policy
Description
TBX3 Polyclonal specifically detects TBX3 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
TBX3 | |
Polyclonal | |
Unconjugated | |
O15119 | |
TBX3 | |
E. coli-derived recombinant human TBX3 Ser321-Thr483 Accession # O15119 | |
100 μg | |
Primary | |
Detects human TBX3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) TBX2, rhTBX6, and rhTBX20 is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1-2 ug/mL, Immunocytochemistry 5-15 ug/mL | |
bladder cancer related protein XHL, T-box 3, T-box protein 3, T-box transcription factor TBX3, TBX3-ISO, ulnar mammary syndrome, UMS, XHL | |
Goat | |
Affinity Purified | |
RUO | |
6926 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction