Learn More
Abnova™ Human TCEAL1 Partial ORF (NP_001006640.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This gene encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. The encoded protein is similar to transcription elongation factor A/transcription factor SII and contains a zinc finger-like motif as well as a sequence related to the transcription factor SII Pol II-binding region. It may exert its effects via protein-protein interactions with other transcriptional regulators rather than via direct binding of DNA. Multiple family members are located on the X chromosome. Alternative splicing results in multiple transcript variants encoding a single isoform. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_001006640.1 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9338 |
Molecular Weight (g/mol) | 36.74kDa |
Name | TCEAL1 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 μg |
Immunogen | MDKPRKENEEEPQSAPKTDEERPPVEHSPEKQSPEEQSSEEQSSEEEFFPEELLPELLPEMLLSEERPPQEGLSRKDLFEGRPPMEQPPCGVGKHKLEEG |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.