Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TCEB3 Partial ORF (NP_003189, 81 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89964265 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-964-265 10 ug
89-964-266 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-964-265 Supplier Abnova™ Supplier No. H00006924Q01S

Used for AP, Array, ELISA, WB-Re

This gene encodes the protein elongin A, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation. [provided by RefSeq]

Sequence: NAEPDEQDFEKSNSRKRPRDALQKEEEMEGDYQETWKATGSRSYSPDHRQKKHRKLSELERPHKVSHGHERRDERKRCHRMSPTYSSDPESSDYGHVQSPPSCTSPHQMY

Specifications

Accession Number NP_003189
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6924
Molecular Weight (g/mol) 37.84kDa
Name TCEB3 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen NAEPDEQDFEKSNSRKRPRDALQKEEEMEGDYQETWKATGSRSYSPDHRQKKHRKLSELERPHKVSHGHERRDERKRCHRMSPTYSSDPESSDYGHVQSPPSCTSPHQMY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ38760/FLJ42849/SIII/TCEB3A
Common Name TCEB3
Gene Symbol TCEB3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.