Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TCF-12/HTF4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB69281
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB69281 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB69281 Supplier R&D Systems Supplier No. MAB69281
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TCF-12/HTF4 Monoclonal specifically detects TCF-12/HTF4 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen TCF-12/HTF4
Applications Immunocytochemistry
Classification Monoclonal
Clone 690129
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99081
Gene Alias BHLHB20, bHLHb20E-box-binding protein, Class B basic helix-loop-helix protein 20, HEB, HEBDNA-binding protein HTF4, helix-loop-helix transcription factor 4, HsT17266, HTF4, HTF4TCF-12, TCF12, transcription factor 12, Transcription factor HTF-4
Gene Symbols TCF12
Host Species Mouse
Immunogen E. coli-derived recombinant human TCF-12/HTF4 Met108-Glu521 Accession # Q99081
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6938
Test Specificity Detects human TCF-12/HTF4 in direct ELISAs. In direct ELISAs, 75%-100% cross-reactivity with recombinant human (rh) TCF-3 and no cross-reactivity with rhTCF-1, -2, -4, or rhStathmin-2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.