Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TCF-2/HNF-1 beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3330SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3330SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3330SP Supplier R&D Systems Supplier No. AF3330SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

TCF-2/HNF-1 beta Polyclonal specifically detects TCF-2/HNF-1 beta in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen TCF-2/HNF-1 beta
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P35680
Gene Alias FJHN, hepatocyte nuclear factor 1-beta, HNF-1 beta, HNF1 beta A, HNF1 homeobox B, HNF1B, HNF-1B, HNF1beta, HNF-1-beta, HNF2, Homeoprotein LFB3, LFB3, LF-B3, MODY5, MODY5HPC11, TCF2, TCF-2, TCF2transcription factor 2, hepatic; LF-B3; variant hepatic nuclear factor, Transcription factor 2, transcription factor 2, hepatic, Variant hepatic nuclear factor 1, vHNF1
Gene Symbols HNF1B
Host Species Goat
Immunogen E. coli-derived recombinant human TCF-2/HNF-1 beta Gly51-Gln182 Accession # P35680
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6928
Test Specificity This antibody detects TCF-2/HNF-1 beta in direct ELISAs and Western blots. It is expected to recognize all three TCF-2 isoforms. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) rhTCF-1, rhTCF-3, and rhTCF-4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.