Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TCF-3/E2A Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB44062
This item is not returnable.
View return policy
Description
TCF-3/E2A Monoclonal specifically detects TCF-3/E2A in Human samples. It is validated for Western Blot.Specifications
TCF-3/E2A | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
NP_003191 | |
TCF3 | |
E.coli-derived recombinant human TCF-3/E2A Gln100-Glu500 Accession # NP_003191 | |
100 μg | |
Primary | |
Detects human TCF-3/E2A in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) TCF-1 (aa 116-233), rhTCF-1 (aa 217-358), rhTCF-1 alpha/LEF-1, rhTCF-2, rhTCF-4, rhTCF-12, rhTCF7L1 (aa 151-320), rhTCFL1 (aa 429-581), or rhTCF7L2 (aa 131-300) is observed. | |
Human | |
Purified |
Western Blot | |
401703 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BHLHB21, bHLHb21TCF-3, Class B basic helix-loop-helix protein 21, E2A, E2A immunoglobulin enhancer-binding factor E12/E47, E2AKappa-E2-binding factor, Immunoglobulin enhancer-binding factor E12/E47, Immunoglobulin transcription factor 1, ITF1, ITF1Transcription factor ITF-1, Kappa-E2-binding factor, MGC129647, MGC129648, Pan2, Tcfe2a, Transcription factor 3, transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47), transcription factor E2-alpha, VDIR, VDR interacting repressor | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6929 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction