Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TCL1A Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4847
This item is not returnable.
View return policy
Description
TCL1A Polyclonal specifically detects TCL1A in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
TCL1A | |
Polyclonal | |
Unconjugated | |
P56279 | |
TCL1A | |
E. coli-derived recombinant human TCL1A Met1-Asp114 Accession # P56279 | |
100 μg | |
Primary | |
Detects human TCL1A in Western blots. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1-PEN, TCL1T-cell leukemia/lymphoma protein 1A | |
Goat | |
Affinity Purified | |
RUO | |
8115 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction