Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TDAG8/GPR65 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10077SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10077SP 25 μg
MAB10077100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10077SP Supplier R&D Systems Supplier No. MAB10077SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TDAG8/GPR65 Monoclonal specifically detects TDAG8/GPR65 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen TDAG8/GPR65
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 976128
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL, Immunocytochemistry 8-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q81IYL9
Gene Alias G protein-coupled receptor 65, GPR65, G-protein coupled receptor 65, Psychosine Receptor, T-cell death-associated gene 8 protein, TDAG8hTDAG8
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human TDAG8/GPR65, Met1-Glu337, Accession # Q81IYL9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 8477
Test Specificity Detects human TDAG8/GPR65 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.