Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TDO2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC9768R100UG
This item is not returnable.
View return policy
Description
TDO2 Monoclonal specifically detects TDO2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
TDO2 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
chky, TDO, TDOTO, TO, TPH2EC 1.13.11.11, TRPO, TRPOtryptophanase, Tryptamin 2,3-dioxygenase, tryptophan 2,3-dioxygenase, Tryptophan Oxygenase, Tryptophan Pyrrolase, Tryptophanase | |
E. coli-derived human TDO2, Leu18-Phe388, Accession # P48775 | |
100 μg | |
6999 | |
Human | |
Supernatant |
Flow Cytometry | |
998604 | |
Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
P48775 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human TDO2 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction