Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human TDP-43/TARDBP Recombinant Protein
SDP

Catalog No. NB2228505MG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB2228505MG 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB2228505MG Supplier Novus Biologicals™ Supplier No. NBP2228500.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-414 of Human TDP-43/TARDBP The Recombinant Human TDP-43/TARDBP Protein is derived from E. coli. The Recombinant Human TDP-43/TARDBP Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) Bioactivity
Formulation 20mM Tris-HCl buffer (pH8.0) containing 10% glycerol, 0.4M urea
Molecular Weight (g/mol) 48.8 kDa
Name TARDBP Recombinant Protein
Quantity 0.5 mg
Source E.coli
Storage Requirements Aliquot and store at −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias ALS10, ALS10TAR DNA-binding protein-43
Gene Symbol TARDBP
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Purity or Quality Grade >90% by SDS-PAGE
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.