Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TDRD1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6296SP
This item is not returnable.
View return policy
Description
TDRD1 Polyclonal specifically detects TDRD1 in Human samples. It is validated for Western Blot.Specifications
TDRD1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CT41.1, CT41.1Cancer/testis antigen 41.1, FLJ21082, tudor domain containing 1, tudor domain containing protein 1, tudor domain-containing protein 1 | |
Sheep | |
Affinity Purified | |
RUO | |
56165 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9BXT4 | |
TDRD1 | |
E. coli-derived recombinant human TDRD1 Asn837-Lys968 Accession # Q9BXT4 | |
25 μg | |
Primary | |
Detects human TDRD1 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse TDRD1 is observed and less than 1% cross-reactivity with recombinant human TDRKH is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction